missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Warning: “Hot” Signs

To help manage and sustain clean and safe workplace

Manufacturer:  Brady™ BRD69588

 View more versions of this product

Catalog No. 19-102294



Description

Description

  • Available in wide variety ofsizes and materials

Specifications

Specifications

ANSI Z535, OSHA 1910.145
Black on Orange
Fiberglass
9000 psi
Surface Mount
10 in.
193°F
0.1 in.
14 in.
-40°C to 90°C
-40°F to 193°F
Warning Hot
1 Ea.
Premium Fiberglass
Use in industrial, utility, commercial and institutional environments
25.4 cm
-40°F
Machine and Operational Sign
35.5 cm
SDS
Documents
Product Certifications