Please login to your online account to display your discounted pricing

Brady™ Vial and Tube Labels

$106.20 - $211.37

Specifications

Adhesive Type Acrylic
Adhesive Type Permanent
Color White
Product Type Laboratory Label
For Use With (Application) For use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides
View More Specs
Products
Catalog Number Mfr. No. Height (English) Quantity Width (English) Price Quantity    

19-140-553

 
brady™
B33179492
1 in. 2250 Roll(s) 1 in. Pack of 2250 Each for $196.03

19-140-575

 
brady™
B33155490
0.6 in. 1500 Roll(s) 1.6 in. Pack of 1500 Each for $106.20

19-140-784

 
brady™
B33126490
0.5 in. 2500 Roll(s) 0.9 in. Pack of 2500 Each for $211.37
Description & Specifications

Specifications

Adhesive Type Acrylic
Adhesive Type Permanent
Color White
Product Type Laboratory Label
For Use With (Application) For use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides
For Use With (Equipment) BBP™33 printer