Brady™ Vial Top Labels
Durable Freezerbondz™ Polyester (B-492) Vial Top Labels are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Manufacturer:  Brady™ B33144492
 View more versions of this product
Catalog No. 19-140-765
Description
Description
  • B-490 labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Matte white polyester material has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance of -320 to 212° F (-196 to 100° C)
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Specifications
Specifications
Non-sterile
White
1.27cm
0.5 in. H x 0.5 in. W
Matte
Use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides
1.27cm
3
-196 °C-130 °C
0.081 mm
0.5 in.
0.5 in.
Laboratory Label
Acrylic, Permanent
White
FreezerBondz Labels for the BBP33 Printer
0.5 in. H x 0.5 in. W
0.5 in.
BBP™33
15.24 mm
LabelMark, MarkWare
0.0032 in
15.88 mm
12.7 mm
Polyester
1500 Roll(s)
Documents