Brady™ Tuff-N-Light U-Channel Sign Posts
Tuff-N-Light U-Channel Sign Posts are manufactured from a special composite material making them both lightweight and maintenance-free. Available in closed or open profile styles.
Manufacturer:  Brady™ BRD97207
 View more versions of this product
Catalog No. 19-054-910
Description
Description
  • Tuff-N-Light™ U-Channel Sign Posts are manufactured from a special composite material making them both lightweight and maintenance-free.
  • Available in closed or open profile styles in either white or green.
  • Close profile posts are 6 times torsionally stronger than steel posts and are pointed for easy installation.
  • Open profile posts are for signs up to 18" x 12". Posts are 8' long.
Specifications
Specifications
White
Open Profile
8 ft.
1 Ea.
Open profile posts are for signs up to 18 x 12 in.
U channel sign post
Documents