Brady™ TLS2200™ and TLS-PC™ Vial Labels
TLS2200™ and TLS-PC™ Vial Labels are made of self-laminating vinyl (B-427), an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
Manufacturer:  Brady™ BPTL75461
 View more versions of this product
Catalog No. 29-635-1463
Specifications
Specifications
Acrylic,Permanent
RoHS, Pass 2002/95/EC, DIN VDE 0472/parts 815
1 in.
BMP™71, TLS 2200™, TLS-PC Link™
2.62 in.
Polyester
10000 Pack
Translucent,White
Matte
Cryo vial, laboratory identification
2.54 cm
Label
Self-laminating Label
Documents