Brady™ TLS 2200™/TLS PC Link™ Portable Thermal Transfer Printer Labels for Vial Tops
Labels for vial tops
Manufacturer:  Brady™ PTL98499
 View more versions of this product
Catalog No. 11-877-01
Description
Description
Labels for vial tops
  • Durable, chemical- and solvent-resistant labels; ink will not smear or fade, and the text stays crisp and readable
  • Printable labels allow more information to fit on any size legend
  • Choice of materials suitable for different environments from liquid nitrogen to autoclaves
Specifications
Specifications
Halogen-free (DIN VDE 0472/parts 815), UL
Acrylic,Permanent
Matte
Nylon Cloth
Centrifuge tube, Cryo vial, Eppendorf tube, Laboratory identification, PCR tube, Vial, Vial top
1
Rectangle with Rounded Corners
-196°C to +90°C
12.7 mm
1 in.
0.3 in.
0.3 in.
Surface Mount
White
Label
TLS 2200™/TLS-PC Link™ Label
BMP™71, TLS 2200™, TLS-PC Link™
500/Pk.
-320-194°F
0.165 mm
0.375 x 1 in. (9.5 x 25.4 mm)
25.40 mm
9.53 mm
0.9 cm