Brady™ TLS 2200™/TLS PC Link™ Portable Thermal Transfer Printer Labels for Vial Tops
Labels for vial tops
Manufacturer:  Brady™ PTL83499YL
 View more versions of this product
Catalog No. 11-877-00
Description
Description
Labels for vial tops
  • Durable, chemical- and solvent-resistant labels; ink will not smear or fade, and the text stays crisp and readable
  • Printable labels allow more information to fit on any size legend
  • Choice of materials suitable for different environments from liquid nitrogen to autoclaves
Specifications
Specifications
Halogen-free (DIN VDE 0472/parts 815), UL
Acrylic,Permanent
Matte
Nylon Cloth
Laboratory identification, vial, vial top
500/Pk.
-320-194°F
0.5 in.
Surface Mount
Yellow
Label
TLS 2200™/TLS-PC Link™ Label
BMP™71, TLS 2200™, TLS-PC Link™
Round
-196°C to +90°C
12.7 mm