Brady™ TLS 2200™/TLS PC Link™ Portable Thermal Transfer Printer Labels for Vial Tops
Labels for vial tops
Manufacturer:  Brady™ PTL82499
 View more versions of this product
Catalog No. 29-635-1468
Description
Description
  • Durable, chemical- and solvent-resistant labels; ink will not smear or fade, and the text stays crisp and readable
  • Printable labels allow more information to fit on any size legend
  • Choice of materials suitable for different environments from liquid nitrogen to autoclaves
Specifications
Specifications
TLS 2200™/TLS-PC Link™ Label
White
9.5 mm
Laboratory Identification, Vial, Vial Top
1
-320-194°F
0.165 mm
0.375 in.
500/Pk.
Matte
Acrylic,Permanent
Halogen-free (DIN VDE 0472/parts 815), UL
Surface Mount
BMP™71, TLS 2200™, TLS-PC Link™
Round
-196°C to +90°C
12.7 mm
Nylon Cloth
Label