missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ TLS 2200 and TLS-PC Link™Portable Thermal Transfer Printer Labels for Slides

Labels for slides

Manufacturer:  Brady™ PTL97488

 View more versions of this product

Catalog No. 11-877-70



Description

Description

  • Durable, chemical- and solvent-resistant labels; ink will not smear or fade, and the text stays crisp and readable
  • Printable labels allow more information to fit on any size legend

Specifications

Specifications

TLS 2200™/TLS-PC Link™ Label
White
Surface Mount
Asset tracking, Barcoding, Component ID, Equipment ID, General purpose ID, Rating plates, Slide label, Through hole-top
22.86 mm
Square
70 to 120°C
26.03 mm
250/Pk.
Matte
22.86 mm
Acrylic,Permanent
AGA, CSA, UL
0.9 in.
BMP™71, TLS 2200™, TLS-PC Link™
1
-94-248°F
0.091 mm
Polyester
Label
0.9 in.
SDS
Documents
Product Certifications
Promotions

Promotions