Brady™ TLS2200™ and TLS-PC™ General Purpose Labels
Labels for the TLS 2200™ & TLS-PC Link™ Thermal Transfer Portable Printers in a wide variety of materials to meet your application needs.
Manufacturer:  Brady™ PTL108461
 View more versions of this product
Catalog No. 11-877-157
Description
Description
  • Self-Laminating Polyester (B-461) has a permanent acrylic adhesive, good print smudge resistance, solvent resistance and high/low temperature performance.
  • Glossy white finish with permanent acrylic (B-423) meets requirements of a halogen-free material per DIN VDE 0472 part 815 and has a service temperature of -94 to 230° F (-70 to 110° C).
  • Provided in a self-laminated (B-427) format with a white printable zone and translucent overlaminating area to protect text with durable, permanent acrylic adhesive has outstanding water and oil resistance.
  • The adhesive and cloth backing material (B-427) offers excellent holding power, yet allow for clean removal and repositioning.
  • Sticks to any clean, dry surface and is specially formulated topcoat for excellent print quality.
Specifications
Specifications
General Purpose Label
Translucent,White
TLS 2200/TLS PC Link Labels
Centrifuge tube, Eppendorf tube, Laboratory identification, PCR tube
4.7 mm
Rectangle
0.0019 in.
41.27 mm
750/Pk.
Matte
1.5 in.
Acrylic,Permanent
Halogen-free (DIN VDE 0472/parts 815)/RoHS Pass 2002/95/EC
0.18 in.
BMP™71, TLS 2200™, TLS-PC Link™
1
-196 °C to 110 °C
0.048 mm
Polyester
Label
Non-sterile
38.1 mm