Brady™ TLS PC Link™ Thermal Labeling System
Rugged desktop/portable printer with multiple labeling capabilities
$64.45 - $81.42
Specifications
Quantity Each
View More Specs
Includes: Windows drivers, LabelMark labeling software, rechargeable battery pack, AC power supply/battery charger, R6210 ribbon, sample labels, Quick Start Guide, manuals, and communications cable
Products ${productFamilyLength}
Catalog Number Mfr. No. Certifications/Compliance For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Certifications/Compliance For Use With (Equipment) Price Quantity  
19-150-101 Brady™
MBATT18801
RoHS Brady TLS-PC Link™ thermal labeling system Each for $81.42
19-150-102 Brady™
TLSPCAC
UL, CSA Brady TLS-PC Link™ thermal labeling system Each for $64.45
Specifications
Description
Lightweight desktop/portable printer system has thermal transfer printing, bar coding, and multiple material configuration capabilities.
  • Included Windows™ drivers permit labels to be printed from LabelMark™ software (included), as well as BradySoft™, or any software that is Windows compatible; firmware may be upgraded through the mini-Din serial jack
  • Printer utilizes TLS2200 printer materials and ribbons, plus bulk rolls of material
  • Works with specialty lab materials which are sized to specific vial and tube sizes
  • Withstands tough lab conditions including liquid nitrogen, freezer autoclave, xylene, ethanol and DMSO
  • Prints up to 500 labels between charges
Order Info
Uses AC power or rechargeable battery pack; replacement power supply/battery charger and rechargeable battery pack available separately.
Description & Specifications
Specifications
Each