Brady™ TLS PC Link™ Thermal Labeling System
Rugged desktop/portable printer with multiple labeling capabilities
Manufacturer:  Brady™ TLSPCAC
 View more versions of this product
Catalog No. 19-150-102
Includes: Windows drivers, LabelMark labeling software, rechargeable battery pack, AC power supply/battery charger, R6210 ribbon, sample labels, Quick Start Guide, manuals, and communications cable
Description
Description
Lightweight desktop/portable printer system has thermal transfer printing, bar coding, and multiple material configuration capabilities.
  • Included Windows™ drivers permit labels to be printed from LabelMark™ software (included), as well as BradySoft™, or any software that is Windows compatible; firmware may be upgraded through the mini-Din serial jack
  • Printer utilizes TLS2200 printer materials and ribbons, plus bulk rolls of material
  • Works with specialty lab materials which are sized to specific vial and tube sizes
  • Withstands tough lab conditions including liquid nitrogen, freezer autoclave, xylene, ethanol and DMSO
  • Prints up to 500 labels between charges
Order Info
Uses AC power or rechargeable battery pack; replacement power supply/battery charger and rechargeable battery pack available separately.
Specifications
Specifications
3000mA
Provides an extra power supply for use with the Brady TLS-PC Link™ thermal labeling system
120V at 60Hz
UL, CSA
Brady TLS-PC Link™ thermal labeling system
Each
Documents