Brady™ TLS 2200™ and TLS PC Link™ Printer Ribbons
For use with the TLS2200 and TLS PC Link Thermal Transfer Printers
Manufacturer:  Brady™ R4310
 View more versions of this product
Catalog No. 29-635-1473
Description
Description
Black ribbons for use with a variety of Brady label materials.
R4310 Series Ribbon
Recommended for use with these label materials:
  • Matte white PE tag (B-109)
  • PermaSleeve™ 3:1 heat-shrink polyolefin (B-342)
  • Matte white permanent paper (B-424)
  • High-temperature amber polyamide (B-426)
  • Self-laminating vinyl (B-427)1
  • Matte metallized polyester (B-428)2
  • Self-extinguishing Tedlar™ (B-437)
  • Ultra-aggressive permanent matte metallized polyester (B-486)
  • Matte white (B-489)3
  • Matte white nylon cloth (B-499); and self-laminating Tedlar (B-642)
R4410 and R6010 Series Ribbons
Recommended for use with these permanent labels:
  • lGlassy white (B-423)2,3 (only with 6010 ribbon)
  • Super-permanent polyester (B-422)2
  • Glossy clear polyester (B-430)2
  • Glossy metallized (B-435)2
  • Glossy white vinyl (B-439)
  • Matte white polyester (B-459)2
  • Ultra-aggressive glossy white polyester (B-483)2
And these other label materials:
  • High-temperature glossy white polyamide (B-457)1
  • Glossy white static dissipative polyester (B-473)2 Polyimide (B-477)1
R6210 Series Ribbon
Recommended for use with these label materials:
  • BradySleeve™ heat-shrink polyolefin (B-321)
  • Matte white polypropylene tag/insert (B-412)
  • Self-laminating vinyl (B-427)
Acceptable for use with these label materials:
  • Matte white tamper-evident vinyl (B-351)
  • Glossy white (B-423)
  • Super-permanent polyester (B-422)
  • Glossy clear permanent polyester (B-430
  • Self-extinguishing Tedlar (B-437)
  • Glossy white permanent vinyl (B-439)
  • Glossy white static dissipative polyester (B-473) and polyimide (B-477)
  • Ultra-aggressive permanent glossy white polyster (B-483
  • Matte white nylon cloth (B-499)
  • Self-laminating Tedlar (B-642)
Specifications
Specifications
Ribbon printer
UL, CSA
Use with a variety of Brady label materials
23 m
Ribbon
2 in.
Black
Black 4300 Series Thermal Transfer Printer Ribbon
R4310 Series Printer
1 Ea.
75 ft.
50.80 mm