Brady™ Thermal Transfer Polypropylene Labels
Sheets of printable labels for use in freezers
Manufacturer:  Brady™ THT644915SC
 View more versions of this product
Catalog No. 19-150-422
Description
Description
  • Polypropylene material
  • Matte finish
  • Color: white
Specifications
Specifications
Acrylic, Removable
RoHS 2002/95/EC
Matte
For barcoding, bottles, component ID, dishes, equipment ID, flasks, general lab ID, general purpose ID, laboratory identification
1.9cm
24 lb./in.
1.5 in.
1 in.
Polypropylene
White
2.54cm
0.75 in.
Thermal transfer printer
2 Years
0.0044 in.
38.1mm
Label
1500 Roll(s)
Documents