Brady™ Ten-Lock Lockout Stations
Provide centralized access to lockout/tagout materials
$217.64 - $474.60
Includes: Components: 10 blue bumper padlocks, 25 “Do Not Operate” tags, and three 1 in. (2.5cm) safety lockouts
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
19-016527 Brady™
BRD65679
Each for $217.64
19-016528 Brady™
BRD65680
Each for $474.60
Specifications
Description
  • Mounts on wall or other flat surface
  • Available with or without lockout/tagout components
Order Info
Dimensions (L x W): 19 × 12 in. (48.3 × 30.5cm).