Please login to your online account to display your discounted pricing

Brady™ Signs: “Warning: Step Down”

Provides a strong and noticeable reminder for employees to protect themselves and maintain a safe work atmosphere

$8.18 - $25.31

Specifications

Color Black on Orange
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Message Warning Step Down
Type Fall protection sign
Quantity Each
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W) Material Temperature Range (Metric) Thickness Price Quantity    

19-035-805

 
brady™
BRD69608
Each for $25.31

19-035808

 
brady™
BRD85139
Each for $8.18

19-036-428

 
brady™
BRD85140
Each for $11.49
Description & Specifications

Specifications

Color Black on Orange
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Message Warning Step Down
Type Fall protection sign
Quantity Each
Style Surface Mount

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “WARNING STEP DOWN”
  • Black on orange
  • Designed to withstand harsh industrial environments
  • Available in a variety of sizes and materials
Materials
  • B-401: Plastic — high-impact polystyrene
  • B-120: Premium fiberglass — fiberglass reinforced polyester
  • B-302: Self-sticking polyester — surface printed polyester with polyester overlaminate and permanent pressure-sensitive adhesive