missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Signs: Danger Hydrochloric Acid

“Danger: Hydrochloric Acid” signs are safety signs from Brady™ that are available in a variety of materials, allowing you to choose the material and size that best fits your application.

Manufacturer:  Brady™ BRD25449

 View more versions of this product

Catalog No. 19-102406



Description

Description

  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
Specifications

Specifications

ANSI Z535, OSHA 1910.145, ASTM D 1593
-40°F to 175°F
DANGER Hydrochloric Acid
1 Ea.
Surface Mount
10 in.
-40°F
Chemical and Hazardous Materials Sign
35.56 cm
10 x 14 in.
Black/Red on White
Plastic
Non-adhesive
Light duty industrial, utility, commercial, institutional environments
175°F
0.06 in.
14 in.
-40°C to 80°C
SDS
Documents