missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Self-Laminating Vinyl Wire Markers

For use with BMP™71 Label Maker

Manufacturer:  Brady™ M7164427

 View more versions of this product

Catalog No. 19-150-382



Description

Description

  • Use for wire and cable identification
  • Use in laboratories for vial identification
  • Protected are offers excellent chemical and abrasion resistance
  • Non-printed area wraps around and overlaminated printed text
  • Color: White

Order Info

Additional sizes and colors available. Contact your Fisher Safety Customer Service Representative for ordering information.

Specifications

Specifications

Acrylic, Permanent
RoHS 2005/618/EC, MCV RoHS 2002/95/EC, UL
25.40 mm85.73 mm
Matte
Communications cables, voice/data identification
9.5cm
Rectangle
-40 to 70°C
88.90 mm
25.40 mm
1-0000
Vinyl
100 Roll(s)
White, Translucent
BMP71 Label Printer Labels
1 x 3.38 in.
3.75 in.
28.58 mm
1
-40 to 158°F
0.102 mm
1 in.
25.40 mm
Cartridge
Wire Marker Label
Documents