missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Self-Laminating Vinyl Wire Markers

For use with BMP™71 Label Maker

Manufacturer:  Brady™ M7118427

 View more versions of this product

Catalog No. 19-150-372



Description

Description

  • Use for wire and cable identification
  • Use in laboratories for vial identification
  • Protected are offers excellent chemical and abrasion resistance
  • Non-printed area wraps around and overlaminated printed text
  • Color: White

Order Info

Additional sizes and colors available. Contact your Fisher Safety Customer Service Representative for ordering information.

Specifications

Specifications

Acrylic, Permanent
UL, RoHS 2005/618/EC, MCV RoHS 2002/95/EC
Matte
Panel identification, wire and cable marking
-40 to 158°F
0.75 in.
Vinyl
250 Roll(s)
White, Translucent
0.75 x 1 in.
1 in.
2.54cm
-40 to 70°C
Label
Wire Marker Label
Documents