Brady™ Self-laminating Polyester Laboratory Labels
Self-laminating Polyester Laboratory Labels (B-461) is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
Manufacturer:  Brady™ B33127461
 View more versions of this product
Catalog No. 19-140-651
Description
Description
  • Self-Laminating Polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Service temperature of -320 to 230° F (-196 to 110° C)
Specifications
Specifications
Non-sterile
White, Clear
RoHS
0.5 in. H x 2.2 in. W
Matte
Laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides
19.05 mm
15.24 mm
LabelMark, MarkWare
0.0019 in
61.60 mm
55.88 mm
Polyester
1500 Roll(s)
Acrylic, Permanent
Clear/White
Self-laminating Polyester Labels for the BBP33 Printer
0.5 in. H x 2.2 in. W
2.1 in.
BBP™33
5.3cm
3
-196 °C-110 °C
0.048 mm
0.5 in.
12.70 mm
Laboratory Label
Documents