Please login to your online account to display your discounted pricing

Brady™ Self-Laminating Danger Tags with Different Fronts and Backs

Personalize with special instructions, hazards, or warnings

$34.17 - $34.18

Specifications

Certifications/Compliance ASTM D1593, ASTM D638
Color Black/Red on White
Diameter (English) Hole 0.37 in.
Diameter (Metric) Hole 0.93cm
Material Polyester
View More Specs
Includes:  Nylon fasteners
Products
Catalog Number Mfr. No. Message Price Quantity    

19-040-801

 
brady™
BRD86458
Header: DANGER, Front: DO NOT OPERATE / SIGNED BY / DATE / PHONE, Back: DO NOT REMOVE THIS TAG / Remarks Pack of 10 Each for $34.17

19-040-802

 
brady™
BRD86459
Header: DANGER, Front: OUT OF SERVICE / SIGNED BY / DATE / PHONE, Back: DO NOT REMOVE THIS TAG / Remarks Pack of 10 Each for $34.18
Description & Specifications

Specifications

Certifications/Compliance ASTM D1593, ASTM D638
Color Black/Red on White
Diameter (English) Hole 0.37 in.
Diameter (Metric) Hole 0.93cm
Material Polyester
Message Header: DANGER, Front: DO NOT OPERATE / SIGNED BY / DATE / PHONE, Back: DO NOT REMOVE THIS TAG / Remarks
Durability 3 to 5 Years
For Use With (Application) For marking on-the-spot warning of dangerous conditions
Height (English) 5.75 in.
Height (Metric) 14.6cm
Includes 10 nylon ties
Temperature Range (English) -40° to 248°F
Temperature Range (Metric) -40° to 120°C
Thickness 0.36mm
Width (English) 3 in.
Width (Metric) 7.62cm

  • Protect and seal personalized messages with clear, 1 mil polyester adhesive overlay
  • 0.38 in. (0.95cm) grommeted hole

Dimensions (L x W): 5.75 × 3 in. (14.6 × 7.6cm).