Brady™ Red Striped “Do Not Operate” Lockout Tags
Red Striped “Do Not Operate” economy polyester (B-851) lockout tags are perfect for marking on-the-spot warnings of dangerous conditions.
Manufacturer:  Brady™ BRD66064
Catalog No. 29-635-8302
Description
Description
  • B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
  • Textured write-on surface allows for authorized signatures, dates and other special instructions
  • Withstands moisture, grease and dirt
  • Stays legible longer than cardstock