Brady™ Radiation and Laser Signs
Radiation and Laser safety signs from Brady™ help manage and sustain a clean and safe workplace. They are available in a variety of materials and sizes to best fit your application.
$14.39 - $39.78
Specifications
Quantity 1 Ea.
Min. Temperature (English) -40°F
Product Type Radiation and Laser Warning Sign
View More Specs
Products 7
Catalog Number Mfr. No. Displayed Text Dimensions (L x W) Color Material Price Quantity  
Catalog Number Mfr. No. Displayed Text Dimensions (L x W) Color Material Price Quantity  
19-807-285
View Documents Promotion Details
Brady™
88707
Laser Radiation Avoid Direct Exposure To Beam Class 111B Laser Product 7 x 10 in. Red on White Polyester
Each for $15.57
19-807-287
View Documents Promotion Details
Brady™
89112
CAUTION RADIOACTIVE MATERIAL 2.25 x 2.25 in. Pink on Yellow Polyester
Each for $15.81
19-807-382
View Documents Promotion Details
Brady™
88750
CAUTION RADIOACTIVE MATERIAL 3.5 in. x 10 in. Pink on Yellow Polyester
Each for $14.39
19806988
View Documents Promotion Details
Brady™
25276
CAUTION RADIATION AREA 10 x 7 in. Pink on Yellow Polystyrene
Each for $15.88
19806989
View Documents Promotion Details
Brady™
25277
CAUTION RADIATION AREA 14 x 10 in. Pink on Yellow Polystyrene
Each for $23.34
19-806-990
View Documents Promotion Details
Brady™
42853
CAUTION RADIATION AREA 10 x 7 in. Pink on Yellow Aluminum
Each for $20.57
19-806-991
View Documents Promotion Details
Brady™
BRD42854
CAUTION RADIATION AREA 14 x 10 in. Pink on Yellow Aluminum
Each for $39.78
Description
Description
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Specifications
Specifications
1 Ea.
-40°F
Radiation and Laser Warning Sign
SDS
missing translation for 'documents'
missing translation for 'documents'