missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Production Status Tags
Production Status Tags help you effectively monitor and control production and equipment maintenance.
Supplier: Brady™ 86763
Description
- The excellent write-on surface of B-853 Cardstock is ideal for adding on-the-spot customized notes and other information
- Paper cardstock tags are recommended for indoor use only
- Includes 3/8 in. metal grommet
- Not recommended for chemical exposure
Specifications
Production Status Tag | |
Cardstock | |
0.375 in. | |
3 x 5.75 in. | |
To help you monitor and control production and equipment maintenance | |
76.2 mm | |
−40° to +194°F | |
0.25 mm | |
146 mm |
Black on Yellow | |
ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145 | |
9.52 mm | |
Hazardous Waste Description/Source/Liquid/Solid/Weigth/Volume/Hazard(S)/Toxic/Corrosive/Ignitable/Reactive/By/Date | |
3 in. | |
(100) 12 in. cotton string ties | |
−40° to +90°C | |
5.75 in. |