Please login to your online account to display your discounted pricing

Brady™ PowerMark; Two-Color Printer Ribbons

$267.70 - $319.20

Specifications

Certifications/Compliance RoHS 2005/618/EC, MCV 2002/95/EC
Format Ribbon
Product Type Printer Ribbon
Quantity Each
For Use With (Application) Indoor and outdoor use
View More Specs
Products
Catalog Number Mfr. No. Width (English) Color Price Quantity    

19-041-026

 
brady™
BRD13525
6.25 in. Black Each for $279.97

19-041-027

 
brady™
BRD13520
8.8 in. Black Each for $309.95

19-041-030

 
brady™
BRD13528
8.8 in. Black Each for $309.95

19-041-028

 
brady™
BRD13524
8.8 in. Black Each for $319.20

19-041-029

 
brady™
BRD13526
8.8 in. Black Each for $309.95

19-041-111

 
brady™
BRD13527
6.25 in. Black Each for $279.97

19-041-112

 
brady™
BRD13529
6.25 in. Black Each for $279.97

19-041-113

 
brady™
BRD13522
8.8 in. Black Each for $309.95

19-000-597

 
brady™
13523
6.25 in. Black Each for $267.70
Description & Specifications

Specifications

Certifications/Compliance RoHS 2005/618/EC, MCV 2002/95/EC
Format Ribbon
Product Type Printer Ribbon
Quantity Each
For Use With (Application) Indoor and outdoor use
For Use With (Equipment) BBP™85, PowerMark™ printer
Length (English) 200 ft.
Length (Metric) 60.9m