Please login to your online account to display your discounted pricing

Brady™ PowerMark; Single-Color Printer Ribbons

$239.96 - $269.96

Specifications

Format Ribbon
Product Type Printer Ribbon
Quantity Each
For Use With (Application) For outdoors and in industrial areas
For Use With (Equipment) BBP™85, PowerMark™ printer
View More Specs
Products
Catalog Number Mfr. No. Width (English) Color Price Quantity    

19-041-170

 
brady™
BRD13598
6.25 in. Magenta Each for $239.96

19-041-177

 
brady™
BRD13516
8.8 in. Green Each for $269.96

19-041-179

 
brady™
BRD13518
8.8 in. Yellow Each for $269.96

19-041-180

 
brady™
BRD13519
8.8 in. Orange Each for $269.96

19-041-178

 
brady™
BRD13517
8.8 in. Magenta Each for $269.96
Description & Specifications

Specifications

Format Ribbon
Product Type Printer Ribbon
Quantity Each
For Use With (Application) For outdoors and in industrial areas
For Use With (Equipment) BBP™85, PowerMark™ printer
Length (English) 200 ft.
Length (Metric) 60.9m