Please login to your online account to display your discounted pricing

Brady™ PowerMark; Four-Color Printer Ribbons

$400.00 - $400.00

Specifications

Format Ribbon
Product Type Printer Ribbon
Quantity Each
For Use With (Application) Indoor and outdoor use
For Use With (Equipment) PowerMark™ printer, BBP™85
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    

19-041-033

 
brady™
BRD13530
Black Each for $400.00

19-041-034

 
brady™
BRD13534
Black Each for $400.00

19-041-116

 
brady™
BRD13532
Black Each for $400.00
Description & Specifications

Specifications

Format Ribbon
Product Type Printer Ribbon
Quantity Each
For Use With (Application) Indoor and outdoor use
For Use With (Equipment) PowerMark™ printer, BBP™85
Length (English) 200 ft.
Length (Metric) 60.9m
Width (English) 8.8 in.