Brady™ Polyester Label Cartridges for BMP™21-PLUS and BMP™21-LAB Printers
For use with BMP™21-PLUS and BMP™21-LAB printers
Manufacturer:  Brady™ M21750423
 View more versions of this product
Catalog No. 19-102-741
Description
Description
  • For smooth surface indoor labeling
  • Made of white or clear self laminating polyester material
  • Glossy finish and permanent acrylic adhesive
  • M21-250-423, M21-250-430 and M21-250-430-WT-CL are compatible only with the BMP™21-PLUS printer
Applications:
Asset Tracking, Barcoding, Component ID, Equipment ID, General ID, General Identification, General Purpose ID, Location ID, Outlet/Jack ID, Patch Panel, Patch Panel - ID Port, Plate, Rating Plates, Datacom Equipment
Specifications
Specifications
Label Cartridge
Black,White
Surface Mount
Asset tracking, Barcoding, Component ID, Equipment ID, General ID, General purpose ID, Plate, Rating plates
6.4 cm
Polyester
Cartridge, Label
21 ft.
19.05 mm
Acrylic,Permanent
AGA, CSA, Halogen-free (DIN VDE 0472/parts 815), UL
21 x 0.75 x 21 ft. (6.4 m x 19.05 x 6.4 cm)
BMP™21, ID PAL, LABPAL
6.4 m
1 Roll
Gloss
0.75 in.