missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ “Oxygen: No Smoking...” Signs

“Oxygen: No Smoking...” safety signs from Brady™ help to manage and sustain a clean and safe workplace and are available in a variety of materials to best fit your application.

Manufacturer:  Brady™ BRD25138

 View more versions of this product

Catalog No. 19-102448



Description

Description

  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
Specifications

Specifications

OSHA
-40°F to 175°F
Oxygen No Smoking No Open Flames
1 Ea.
Indoor and outdoor use, light duty industrial, utility, commercial and institutional environment
175°F
0.06 in.
10 in.
-40°C to 80°C
7 x 10 in.
White on Red
Plastic
Non-adhesive
7 in.
-40°F
Chemical and Hazardous Materials Sign
25.4 cm
SDS
Documents