missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Chemical Absorbent Pads

Color-coded absorbents for safe clean-up of hazardous fluids. Brady™ Chemical Absorbent Pads are surfactant treated used on wide range of chemicals.

Manufacturer:  Brady™ BRDCH1212

 View more versions of this product

Catalog No. 19-150-259



Description

Description

  • Made of MAXX technology
  • Benefits of sign and an absorbent in one
  • Highly absorbent for all-purpose industrial applications
  • Ideal for areas where the risk of slips, trips and falls are greatest
  • Chemically inert, do not react with aggressive fluids
  • Perforations provide cost control and reduce waste
  • Medium weight - optimal for most general maintenance needs
Specifications

Specifications

200/Cs.
EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2)
30 gal.
30.48cm
Chemical Absorbent Pad
Medium
7.25kg
30.48cm
12 x 12 in. (30.48 x 30.48cm)
Perforated
Yellow
113.5L
12 in.
Meltblown Polypropylene
16 lb.
12 in.
30 gal., 113.5L
30 gal.
Documents