Please login to your online account to display your discounted pricing

Brady™ Laser and Ink Jet Sign Blanks: Notice

$132.40 - $132.40

Specifications

Adhesive Type Acrylic
Adhesive Type Pressure-sensitive
Color Blue
Color White
Product Type Sign Blank
View More Specs
Products
Catalog Number Mfr. No. Height (English) Width (English) Price Quantity    

19-035-480

 
brady™
BRD12912
3.5 in. 5 in. Pack of 100 Each for $132.40

19-037-455

 
brady™
BRD12913
5 in. 7 in. Pack of 50 Each for $132.40

19-102668

 
brady™
BRD12914
7 in. 10 in. Pack of 25 Each for $132.40
Description & Specifications

Specifications

Adhesive Type Acrylic
Adhesive Type Pressure-sensitive
Color Blue
Color White
Product Type Sign Blank
Display Text Notice
For Use With (Application) Used for safety, maintenance and general markings within a facility
For Use With (Equipment) Laser and inkjet printer
Material Polyester
Quantity 25/pk

Use these blanks and your PC to quickly design professional signs and tags using most Windows™ inkjet/laser printers. (Brady™ MarkWare™ Facility Identification Software required.) Sign and tag blanks may be ordered separately, in a variety of sizes and header designations.