missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Laser and Ink Jet Sign Blanks: Notice

Laser and Ink Jet Notice Blank Signs are made of laser printable Permanent Polyester (B-747) and are recommended for alphanumeric and barcode applications and work well on the topside of most circuit boards and ICs.

Manufacturer:  Brady™ BRD12914

 View more versions of this product

Catalog No. 19-102668



Description

Description

  • Pre-printed headers result in multicolor header signs using a single, black print pass
  • Polyester film with an acrylic pressure sensitive adhesive and a printable coating formulated specially for laser printers and toners
  • High resolution, good solvent and smudge resistance, moderate to high temperature performance
  • White is a UL recognized component and CSA accepted material when printed with designated laser printers
Specifications

Specifications

Sign Blank
Blue,White
7 in.
Laser and inkjet printer
Polyester
7 x 10 in.
10 in.
Acrylic,Pressure-sensitive
Notice
Used for safety, maintenance and general markings within a facility
17.78 cm
25/Pk.
Non-sterile
254 mm
SDS
Documents
Product Certifications