Please login to your online account to display your discounted pricing

Brady™ Laser and Ink Jet Sign Blanks: Danger

$132.40 - $132.40

Specifications

Adhesive Type Acrylic
Adhesive Type Pressure-sensitive
Color Black
Color Red
Color White
View More Specs
Products
Catalog Number Mfr. No. Height (English) Product Type Quantity Width (English) Price Quantity    

19-035-453

 
brady™
BRD12906
3.5 in. Sign Blank 25 Pack 5 in. Pack of 100 Each for $132.40

19-035455

 
brady™
BRD12907
5 in. Sign Blank 25 Pack 7 in. Pack of 25 Each for $132.40

19-102666

 
brady™
BRD12908
7 in. Sign Blank 25 Pack 10 in. Pack of 25 Each for $132.40

19-102670

 
brady™
BRD12917
Each for $132.40
Description & Specifications

Specifications

Adhesive Type Acrylic
Adhesive Type Pressure-sensitive
Color Black
Color Red
Color White
Display Text Danger
For Use With (Application) Used for safety, maintenance and general markings within a facility
For Use With (Equipment) Laser and inkjet printer
Material Polyester

Use these blanks and your PC to quickly design professional signs and tags using most Windows™ inkjet/laser printers. (Brady™ MarkWare™ Facility Identification Software required.) Sign and tag blanks may be ordered separately, in a variety of sizes and header designations.