Please login to your online account to display your discounted pricing

Brady™ Laser and Ink Jet Sign Blanks: Caution

$132.40 - $132.40

Specifications

Adhesive Type Acrylic
Adhesive Type Pressure-sensitive
Color Black
Color White
Color Yellow
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

19-036-514

 
brady™
BRD12910
Pack of 50 Each for $132.40

19-036-515

 
brady™
BRD12909
Pack of 100 Each for $132.40

19-102667

 
brady™
BRD12911
Pack of 25 Each for $132.40
Description & Specifications

Specifications

Adhesive Type Acrylic
Adhesive Type Pressure-sensitive
Color Black
Color White
Color Yellow
Product Type Sign Blank
Display Text Caution
For Use With (Application) Used for safety, maintenance and general markings within a facility
For Use With (Equipment) Laser and inkjet printer
Material Polyester

Use these blanks and your PC to quickly design professional signs and tags using most Windows™ inkjet/laser printers. (Brady™ MarkWare™ Facility Identification Software required.) Sign and tag blanks may be ordered separately, in a variety of sizes and header designations.