Please login to your online account to display your discounted pricing

Brady™ LABXPERT™ Vinyl Cloth General Lab ID Labels

$62.51 - $69.79

Specifications

Certifications/Compliance RoHS
Adhesive Type Repositionable
Adhesive Type Rubber
Color Black
Color White
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Height (English) Height (Metric) Quantity Price Quantity    

11-878-115

 
brady™
X131498
Cryo vial, laboratory identification 0.5 in. 1.2cm 225 Roll(s) Each for $69.79

11-878-119

 
brady™
X78498
Barcoding, bottles, component ID, conical tube, equipment ID, general identification, general purpose ID, petri dish, tube, wire and cable marking 1.9 in. 4.8cm 125 Pack Each for $62.51
Description & Specifications

Specifications

Certifications/Compliance RoHS
Adhesive Type Repositionable
Adhesive Type Rubber
Color Black
Color White
Format Label
Product Type Printer Label
Finish Semi-Gloss
For Use With (Equipment) iDXPERT™, LabXpert™ printer
Material Vinyl Cloth
Description Surface Mount
Width (English) 1 in.

  • Withstand freezers to -80°C (-112°F) and autoclaves to +121°C (+250°F)
  • Semi-gloss white with removable/repositionable adhesive
  • Aggressive tack makes it ideal for use on glass flasks, plastic jugs, bottles, goblets, rough surfaces and general-purpose lab labeling