Please login to your online account to display your discounted pricing
Brady™ LABXPERT™ Printer Labels
Specialty laboratory labels designed for use with these specific label printers
$46.64 - $90.86
Specifications
Adhesive Type Permanent Acrylic
Product Type Laboratory Label
For Use With (Equipment) LabXpert™ printer
View More Specs
Products
Catalog Number Mfr. No. Height (Metric) Material Width (English) Quantity Price Quantity    
22-500-743
brady™
X113490*
5.58cm FreezerBondz™ Polyester 1.5 in. 175 Roll(s) Pack of 175 Each for $90.86
22-500-745
brady™
BRD60907
4.19cm FreezerBondz™ Polyester 0.5 in. 225 Roll(s) Pack of 225 Each for $70.07
22-500-744
brady™
BRD60906
9.52cm FreezerBondz™ Polyester 1.5 in. 100 Roll(s) Pack of 100 Each for $65.69
22-500-754
brady canada
BRD60932
0.47cm Polyester 1.5 in. 450 Roll(s) Pack of 225 Each for $54.42
22-500-758
brady™
BRD60939
5.71cm Polyester 1.5 in. 175 Roll(s) Pack of 175 Each for $74.31
22-500-755
brady canada
BRD60933
9.52cm Polyester 1.5 in. 100 Roll(s) Pack of 500 Each for $49.32
22-500-759
brady™
BRD60940
1.27cm Polyester 0.5 in. 225 Roll(s) Pack of 225 Each for $58.78
22-500-756
brady canada
BRD60935
0.95cm Polyester 1.125 in. 450 Roll(s) Pack of 500 Each for $66.90
22-500-757
brady™
BRD60938
1.52cm Polyester 1.8 in. 225 Roll(s) Pack of 225 Each for $62.21
22-500-764
brady™
BRD60928
3.17cm Polyester 19 in. 1 Roll(s) Each for $60.49
22-500-763
brady™
BRD60927
2.54cm Polyester 19 in. 1 Roll(s) Each for $58.78
22-500-765
brady™
60930
0.95cm Polyester 19 in. 1 Roll(s) Each for $56.33
22-500-771
brady canada
BRD60945
2.54cm Vinyl 1.75 in. 175 Roll(s) Pack of 175 Each for $51.82
22-500-772
brady canada
BRD60946
3.81cm Vinyl 1.75 in. 175 Roll(s) Pack of 175 Each for $69.16
22-500-773
brady™
BRD60947
0.63cm Vinyl 1.1 in. 500 Roll(s) Pack of 500 Each for $46.64
22-500-781
brady™
BRD60937
4.44cm Polyester 1 in. 1 Roll(s) Pack of 225 Each for $62.21
22-500-795
brady canada
60942
1.27cm Polyester 2.2 in. 175 Roll(s) Pack of 225 Each for $65.12
22-500-796
brady™
BRD60944
1.52cm Polyester 2.62 in. 150 Roll(s) Pack of 150 Each for $60.14
Description & Specifications
Specifications
Adhesive Type Permanent Acrylic
Product Type Laboratory Label
For Use With (Equipment) LabXpert™ printer
  • Printable, self-laminating high-performance laboratory identification for LABXPERT printer.
  • Polyester label withstands temps -196° to +121°C
Ultrathin construction prevents jamming in racks & centrifuge