Please login to your online account to display your discounted pricing

Brady™ LABXPERT™ and IDXPERT™ Nylon Cloth Printer Labels

Use these labels for a variety of general labeling needs. Brady™ LABXPERT™ and IDXPERT™ Nylon Cloth Printer Labels are ideal for LABXPERT™ and IDXPERT™ printers.

$48.97 - $79.98

Specifications

Certifications/Compliance UL
Adhesive Type Permanent Acrylic
Format Cartridge, Label
Product Type Printable Label
Finish Matte
View More Specs
Products
Catalog Number Mfr. No. Height (Metric) Shape Width (Metric) Price Quantity    

22-500-748

 
brady™
BRD60911
20.96cm Rectangular 9.53mm Pack of 175 Each for $48.97

22-500-750

 
brady™
BRD60913
12.70mm Rectangular 25.40mm Pack of 225 Each for $71.16

22-500-753

 
brady™
BRD60920
4.8cm Rectangular Pack of 150 Each for $62.55

22-500-760

 
brady™
BRD60948
12.70mm Rectangle and circle 25.40mm Pack of 300 Each for $78.81

22-500-761

 
brady™
BRD60949
12.70mm Rectangular, round 25.40mm Pack of 300 Each for $79.98

22-500-762

 
brady™
BRD60950
9.53mm Rectangle and circle 25.40mm Pack of 300 Each for $78.24
Description & Specifications

Specifications

Certifications/Compliance UL
Adhesive Type Permanent Acrylic
Format Cartridge, Label
Product Type Printable Label
Finish Matte
For Use With (Equipment) iDXPERT™, LabXpert™ printer
Material Nylon Cloth
Shape Rectangular
Style Powder Coated, Smooth, Textured/Rough

  • Nylon cloth labels
  • Matte finish
  • Permanent acrylic adhesive