Brady™ LABXPERT™ and IDXPERT™ Nylon Cloth Printer Labels: Circular
Specialty laboratory labels designed for use with these specific label printers
Manufacturer:  Brady™ X83499ORBK
 View more versions of this product
Catalog No. 29-635-5296
Specifications
Specifications
Cartridge, Label
Black,Orange
1.2 cm
Matte
General and industrial identification, laboratory identification, vial, vial top
1.2 cm
0.5 in.
Nylon Cloth
300 Roll(s)
Acrylic,Permanent
UL
Powder Coated, Smooth, Textured/Rough
0.5 in.
iDXPERT™, LabXpert™ printer
Round
0.5 in.
Printable Label