Brady™ LABXPERT™ and IDXPERT™ Nylon Cloth Printer Labels: Circular
Specialty laboratory labels designed for use with these specific label printers
Manufacturer:  Brady™ X82499RDBK
 View more versions of this product
Catalog No. 29-635-5294
Description
Description
Specifications
Specifications
Printable Label
Black,Red
0.95 cm
0.3 in.
iDXPERT™, LabXpert™ Printer
Round
Nylon Cloth
Cartridge, Label
0.3 in.
Acrylic,Permanent
UL
Powder Coated, Smooth, Textured/Rough
General and industrial identification, Laboratory identification, Vial, Vial top
0.9 cm
0.37 in.
300/Pk.
Matte