Brady™ LABXPERT™ Handheld Labeler
Increase labeling efficiency in your lab
$56.18 - $56.18
Specifications
For Use With For labxpert label maker
View More Specs
Includes: Printer, AC power supply†, PC cable, supply cartridge, and CD with manuals and driver.
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
11-878-103 Brady™
XPERTAC
AC Adapter Each for $56.18
Specifications
Description
Produce professional-looking labels for your lab samples and drastically reduce labeling time. Label vials, vial tops, slides, well plates and more, quickly and easily with durable materials designed specifically for the lab environment.
  • Labels will not fall off or smear, can fit more information, and are always legible
  • 300dpi print head for crisp, clear printing even on the smallest labels using the smallest fonts—from 1 to 32mm
  • Auto-serialization lets you produce 40 labels in less than two minutes—80% faster than handwriting
  • One-touch time and date stamping
  • Barcoding—2D and linear (code 39 and 128)
  • Over 140 lab-specific Greek, research, and safety symbols
  • PC connectivity to download print jobs
  • Drop, Lock & Go Smart Cartridge combines labels and ribbon for a quick and easy change
  • Optional IdentiLab™ Software enables you to create labels on your PC and download database information in three simple steps
  • Easy interface with spreadsheets and many other programs
  • Increase your text options with more fonts, symbols and sizes
  • Work in an easy “click and drag” environment
Order Info
“AA” battery or AC power operation. Variety of LABXPERT labels for numerous lab applications are available separately.
† International model includes international AC power supply.
Description & Specifications
Specifications
For labxpert label maker