Brady™ LABXPERT™ FreezerBondz Labels
For Brady TLS 2200 and PC Link Printers
Manufacturer:  Brady™ X113490
 View more versions of this product
Catalog No. 11-878-109
Description
Description
  • Withstand liquid nitrogen to -196°C (-321°F) , freezer to -80°C (-112°F), and autoclave to +121°C (+250°F)
  • Matte white with permanent acrylic adhesive
  • Will adhere to a frozen or frosted surface without the need to scrape or dry the vial before placement of the label
  • Suitable for glass and polypropylene surfaces
To be assured that label stays secured when placed back into storage, label should wrap around the vial and slightly overlap itself.
Specifications
Specifications
Label
Black,White
Surface Mount
Use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides
5.58 cm
175/Pk.
Matte
Acrylic,Permanent
RoHS 2002/618/EC, MCV RoHS 2002895/EC
2.2 in.
LabXpert™ Printer
Polyester
Label
1.5 in.