Brady™ LABXPERT™ FreezerBondz Labels
For Brady TLS 2200 and PC Link Printers
$133.57 - $156.24
Specifications
Certifications/Compliance RoHS 2002/618/EC, MCV RoHS 2002895/EC
Adhesive Type Acrylic,Permanent
Color Black,White
Format Label
Product Type Label
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Height (English) Height (Metric) Width (English) Price Quantity  
Catalog Number Mfr. No. Quantity Height (English) Height (Metric) Width (English) Price Quantity  
11-878-110
View Documents Promotion Details
Brady™
X114490
100/Roll 3.75 in. 9.52 cm 1.5 in.
Each for $133.57
11-878-108
View Documents Promotion Details
Brady™
X112490
450/Pk. 0.37 in. 0.95 cm 1.25 in.
Each for $156.24
11-878-111
View Documents Promotion Details
Brady™
X124490
225/Pk. 1.6 in. 4 cm 0.5 in.
Each for $137.72
Description
Description
  • Withstand liquid nitrogen to -196°C (-321°F) , freezer to -80°C (-112°F), and autoclave to +121°C (+250°F)
  • Matte white with permanent acrylic adhesive
  • Will adhere to a frozen or frosted surface without the need to scrape or dry the vial before placement of the label
  • Suitable for glass and polypropylene surfaces
To be assured that label stays secured when placed back into storage, label should wrap around the vial and slightly overlap itself.
Specifications
Specifications
RoHS 2002/618/EC, MCV RoHS 2002895/EC
Black,White
Label
Use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides
Polyester
Acrylic,Permanent
Label
Matte
LabXpert™ Printer
Surface Mount