Brady™ Labels for BMP™21 Mobile Printer, ID PAL™ and LABPAL™ Printers
For general identification, laboratory identification, panel identification and plate
Manufacturer:  Brady™ M215007425
 View more versions of this product
Catalog No. 19-140-449
Description
Description
  • Made of B-7425 polypropylene or B-595 vinyl film with pressure sensitive, permanent adhesive
  • Various colors available
<
Compliance
Compliant with RoHS Directive
Specifications
Specifications
Acrylic
RoHS
0.5 in.
BMP™21 mobile printer, ID PAL and LABPAL printer
-112 to 158°F
0.004 in.
Polypropylene
2000 Roll(s)
White
Matte
For general identification, laboratory identification, panel identification and plate
1.27cm
-80 to 70°C
21 ft.
Printer Label