Brady™ Label Maker Cartridges: Indoor/Outdoor Vinyl
Label Maker Cartridges: Vinyl are made of durable, low-shrink vinyl with ultra-aggressive adhesive that are ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
Manufacturer:  Brady™ MC750595ORBK
 View more versions of this product
Catalog No. 19-140-413
Description
Description
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
  • All-in-one cartridge includes ribbon and label supply
Specifications
Specifications
Non-sterile
Black, Orange
50 Gardner Units
25 ft.
BBP™31 sign and label printer, GlobalMark™ industrial label maker, MiniMark™ industrial label printer, PowerMark™ sign and label maker, Labelizer™ Plus sign and labe
8 to 10 Years
-40 to 82°C
0.75 in.
1 Roll(s)
Permanent
ASTM D 523, PSTC-133, RoHS
Gloss
Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels for indoor/outdoor usage
762cm
-40 to 180°F
0.004 in
Vinyl, indoor/outdoor
Documents