Brady™ Label Maker Cartridge: Nylon Cloth, Round
Label Maker Cartridge: Round Nylon Cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
Manufacturer:  Brady™ M83499RDBK
 View more versions of this product
Catalog No. 19-140-395
Description
Description
  • Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable
  • Recommended for indoor use
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • All-in-one cartridge includes label supply and R4300 Series ribbon
Specifications
Specifications
Non-sterile
Acrylic, Permanent
UL, CSA, DIN VDE 0472 Part 815, ASTM D
0.5 in. Dia
Matte White
BMP™51 and BMP™53 printer
-40 to 90°C
0.0065 in
Nylon Cloth
Polyamide
Black, Red
0.95cm
0.5 in. Dia
Wiremarking and general labeling applications
-40 to 194°F
80 lb./in.
0.37 in.
1 Roll(s)
Documents