Brady™ Keyed-Different Safety Locks
Keyed-Different Safety Lock padlocks from Brady™ have nonconductive fiberglass-reinforced nylon bodies and unique key cylinders that prevent electricity from traveling from the shackle to the key.
Manufacturer:  Brady™ 118929
 View more versions of this product
Catalog No. 19-140-339
Description
Description
  • Extremely compact and lightweight - compact safety padlocks from Brady™ are the smallest safety locks on the market
  • These lightweight padlocks comply with all OSHA requirements and offer superior temperature, chemical and corrosion-resistance
  • Non-conductive safety padlocks are a preferred alternative to metal locks for lockout applications
  • Each padlock is keyed differently with keys precision-machined to exacting standards, so that the key for one lock will not open another in the set
Specifications
Specifications
RoHs, OSHA 29 CFR 1910.147, ANSI Standard Z244
1.31 in.
0.2 in.
0.5cm
Compact Safety Padlock
Plastic
1.4 in.
3.6cm
1.2 in.
3.33cm
0.625 in.
1.59cm
Blue
Different
Ideal for electrical contractors/maintenance/electricians
Electrical; Confined Spaces
1 key, 3 language labels/lock
3cm
Documents