Brady™ Keyed-Alike Safety Locks
Keyed-Alike Safety Lock padlocks from Brady™ have nonconductive fiberglass-reinforced nylon bodies and unique key cylinders that prevent electricity from traveling from the shackle to the key.
Manufacturer:  Brady™ BRD118957
 View more versions of this product
Catalog No. 19-140-361
Description
Description
  • Extremely compact and lightweight - compact safety padlocks from Brady™ are the smallest safety locks on the market
  • These lightweight padlocks comply with all OSHA requirements and offer superior temperature, chemical and corrosion-resistance
  • Non-conductive safety padlocks are a preferred alternative to metal locks for lockout applications
  • Every lock in this set can be opened with the same key, which is beneficial when multiple locks are assigned to a single employee
Specifications
Specifications
RoHs, ANSI Z244, OSHA 29 CFR 1910.147
1 in.
1.52cm
0.25 in.
Compact Safety Padlock
Plastic
1.4 in.
3.6cm
1.2 in.
2.54cm
0.6 in.
0.6cm
Orange
Alike
For certain lockout applications
Electrical; Confined Spaces
Key and English, Spanish and French labels, special 5-pin cylinder
3cm
Documents