Please login to your online account to display your discounted pricing
Brady™ IP™ Thermal Transfer Printers
Fully integrated, easy-to-use printer, material, and software system
Manufacturer: brady™  BPIP300
 View more versions of this product
Catalog No. 11-878-504
  • / Each
Description & Specifications
Specifications
Warranty 5 Year(s)
Display LED
Format Printer
For Use With (Application) Barcode labeling, circuit board labeling, control panel/push button, data communications labeling, DoD UID, laboratory identification, product identification, wire and cable marking, wire and cable marking - ADM
Memory 64MB
Power Supply Details AC power and USB, serial and ethernet 10/100 connectivity
Quantity Each
Resolution 300 dpi
Color Capability High Volume
Weight (English) 13 lb.
Weight (Metric) 5.89kg