Brady™ IP™ Thermal Transfer Printers
Fully integrated, easy-to-use printer, material, and software system
$2,697.24 - $3,998.15
Specifications
Display LED
For Use With (Application) Barcode labeling, circuit board labeling, control panel/push button, data communications labeling, DoD UID, laboratory identification, product identification, wire and cable marking, wire and cable marking - ADM
Memory 64MB
Power Supply Details AC power and USB, serial and ethernet 10/100 connectivity
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Capability Resolution Price Quantity  
Catalog Number Mfr. No. Color Capability Resolution Price Quantity  
11-878-504 Brady™
BPIP300
High Volume 300dpi Each for $2,697.24
22-500-845 Brady™
103958
Standard Volume 600dpi Each for $3,998.15
22-500-844 Brady™
103957
Standard Volume 300dpi Each for $2,775.49
Specifications
Description
Smart Cell technology allows the printer, materials, and ribbon to communicate with each other. System offers material recognition, automatic formatting, and user-friendly printer features for labels, sleeves, and tags.
  • High-volume, peripheral-desktop design
  • Produce basic wraps, self-laminating heat-shrink sleeves, and flag-style wire markers
  • Faster print speed and “multiple-up” labels and push-button labels
  • Print Resolution: 300 or 600dpi
  • Print Speed: up to 8 in. (20.3cm) per sec.
  • Connectivity Options: Serial, USB 2.0 and Ethernet 10/100
Description & Specifications
Specifications
LED
Barcode labeling, circuit board labeling, control panel/push button, data communications labeling, DoD UID, laboratory identification, product identification, wire and cable marking, wire and cable marking - ADM
64MB
AC power and USB, serial and ethernet 10/100 connectivity