Brady™ IP™ 6400 Series Thermal Transfer Printer Ribbon
High density ribbon with excellent solvent, high heat and super scratch resistance
Manufacturer:  Brady™ BRD112648
 View more versions of this product
Catalog No. 11-878-521
Specifications
Description
Ribbon, Printer; Brady Identification Solutions; 6400 Series; Thermal transfer; Black; 4 in. x 242 ft. (101.600mm x 73.760m); 0.50 in. core I.D.; For use with BBP11
  • Resin Formulation
  • Black color
Compatible with wide range of printers:
  • IP300, IP600 (F cat.11-878-501)
  • BBP72, BBP81, Brady 200MVP Plus, Brady 2461, Brady 300MVP Plus, Brady 300X-Plus II, Brady 3481, Brady 360X-Plus II, Brady 600X-Plus II, Brady 6441, Bradyprinter PR Plus, Bradyprinter PR Plus 600, PAM 3000, Tagus T200, Tagus T300, Thermal Transfer Printers (F cat. 22-500-855)
  • BBP11 (F cat.11-878-521)
Description & Specifications
Specifications
Black
BBP™11 Label printer
73.8m
0.5 in.
Each
1.27cm
242 ft.
4 in.
Resin